2-Ethoxy-6-methoxyphenylboronic acid 98%
Proveedor: Apollo Scientific
OR907393-10G
OR907393-1G
OR907393-5G
APOSOR907393-10GEA
754
EUR
APOSOR907393-10G
APOSOR907393-1G
APOSOR907393-5G
2-Ethoxy-6-methoxyphenylboronic acid 98%
2-Ethoxy-6-methoxyphenylboronic acid
Formula:
C₉H₁₃BO₄ MW: 196,008 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD28024840 Núm. CAS: 2377606-71-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...