N-(3-Cyanophenyl)cyclopropanecarboxamide 95%
Proveedor: Apollo Scientific
OR908089-1G
OR908089-5G
APOSOR908089-1GEA
190
EUR
APOSOR908089-1G
APOSOR908089-5G
N-(3-Cyanophenyl)cyclopropanecarboxamide 95%
N-(3-Cyanophenyl)cyclopropanecarboxamide
Formula:
C₁₁H₁₀N₂O MW: 186.21 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD03159449 Núm. CAS: 101946-36-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...