Methyl 5-cyano-2-methoxybenzoate 96%

Proveedor: Apollo Scientific

OR908117-1G OR908117-250MG
APOSOR908117-1GEA 273 EUR
APOSOR908117-1G APOSOR908117-250MG
Methyl 5-cyano-2-methoxybenzoate 96%
Methyl 5-cyano-2-methoxybenzoate
Formula: C₁₀H₉NO₃
MW: 191,183 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD11044990
Núm. CAS: 40757-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR