Neopentyl 2-chloro-2-oxoacetate 95%
Proveedor: Apollo Scientific
OR908230-1G
OR908230-5G
APOSOR908230-1GEA
183
EUR
APOSOR908230-1G
APOSOR908230-5G
Neopentyl 2-chloro-2-oxoacetate 95%
Neopentyl 2-chloro-2-oxoacetate
Formula:
C₇H₁₁ClO₃ MW: 178.613 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD17214254 Núm. CAS: 209848-82-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...