(3-Cyclohexylpropyl)boronic acid 95%
Proveedor: Apollo Scientific
OR908380-1G
OR908380-5G
APOSOR908380-1GEA
203
EUR
APOSOR908380-1G
APOSOR908380-5G
(3-Cyclohexylpropyl)boronic acid 95%
(3-Cyclohexylpropyl)boronic acid
Formula:
C₉H₁₉BO₂ MW: 170,057 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD06212395 Núm. CAS: 1498314-72-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...