3-Ethyl-4-methyl-1H-pyrazole hydrochloride 98%

Proveedor: Apollo Scientific

OR909287-1G OR909287-5G
APOSOR909287-1GEA 183 EUR
APOSOR909287-1G APOSOR909287-5G
3-Ethyl-4-methyl-1H-pyrazole hydrochloride 98%
3-Ethyl-4-methyl-1H-pyrazole hydrochloride
Formula: C₆H₁₀N₂·ClH
MW: 146,618 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD18803465
Núm. CAS: 1218791-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR