4-(2-Oxopyrrolidin-1-yl)benzonitrile 95%
Proveedor: Apollo Scientific
OR909556-1G
OR909556-25G
OR909556-5G
APOSOR909556-1GEA
145
EUR
APOSOR909556-1G
APOSOR909556-25G
APOSOR909556-5G
4-(2-Oxopyrrolidin-1-yl)benzonitrile 95%
4-(2-Oxopyrrolidin-1-yl)benzonitrile
Formula:
C₁₁H₁₀N₂O MW: 186,21 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD07345520 Núm. CAS: 167833-93-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...