1-(Methoxymethyl)benzoimidazole 95%
Proveedor: Apollo Scientific
OR910190-1G
OR910190-5G
APOSOR910190-1GEA
219
EUR
APOSOR910190-1G
APOSOR910190-5G
1-(Methoxymethyl)benzoimidazole 95%
1-(Methoxymethyl)benzoimidazole
Formula:
C₉H₁₀N₂O Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00956939 Núm. CAS: 18249-98-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...