1-(3-Methylbutyl)pyrazole 98%
Proveedor: Apollo Scientific
OR910247-10G
OR910247-1G
OR910247-5G
APOSOR910247-10GEA
538
EUR
APOSOR910247-10G
APOSOR910247-1G
APOSOR910247-5G
1-(3-Methylbutyl)pyrazole 98%
1-(3-Methylbutyl)pyrazole
Formula:
C₈H₁₄N₂ MW: 138,21 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD12405510 Núm. CAS: 847818-51-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...