N-tert-Butyl-3-nitropyridin-2-amine 98%
Proveedor: Apollo Scientific
OR911063-1G
OR911063-5G
APOSOR911063-1GEA
168
EUR
APOSOR911063-1G
APOSOR911063-5G
N-tert-Butyl-3-nitropyridin-2-amine 98%
N-tert-Butyl-3-nitropyridin-2-amine
Formula:
C₉H₁₃N₃O₂ MW: 195,218 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00499524 Núm. CAS: 79371-45-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...