Methyl oxindole-4-carboxylate 95%
Proveedor: Apollo Scientific
OR913340-1G
OR913340-250MG
APOSOR913340-1GEA
151
EUR
APOSOR913340-1G
APOSOR913340-250MG
Methyl oxindole-4-carboxylate 95%
Methyl oxindole-4-carboxylate
Formula:
C₁₀H₉NO₃ MW: 191,183 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD08277245 Núm. CAS: 90924-46-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...