(2-Acetyl-phenoxy)-acetic acid 95%
Proveedor: Apollo Scientific
OR913438-1G
APOSOR913438-1GEA
142
EUR
APOSOR913438-1G
(2-Acetyl-phenoxy)-acetic acid 95%
(2-Acetyl-phenoxy)-acetic acid
Formula:
C₁₀H₁₀O₄ MW: 194,184 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00118956 Núm. CAS: 1878-62-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...