(Pyrrol-3-yl)-acetic acid 95%
Proveedor: Apollo Scientific
OR913592-1G
OR913592-250MG
APOSOR913592-1GEA
683
EUR
APOSOR913592-1G
APOSOR913592-250MG
(Pyrrol-3-yl)-acetic acid 95%
(Pyrrol-3-yl)-acetic acid
Formula:
C₆H₇NO₂ MW: 125,125 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD09800534 Núm. CAS: 86688-96-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...