2-Amino-6-chloronicotinonitrile 95%
Proveedor: Apollo Scientific
OR916978-1G
OR916978-250MG
APOSOR916978-1GEA
549
EUR
APOSOR916978-1G
APOSOR916978-250MG
2-Amino-6-chloronicotinonitrile 95%
2-Amino-6-chloronicotinonitrile
Formula:
C₆H₄ClN₃ MW: 153,569 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD18261104 Núm. CAS: 52471-07-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...