7-Methoxyquinoline hydrochloride 95%
Proveedor: Apollo Scientific
OR917812-1G
OR917812-5G
APOSOR917812-1GEA
71.2
EUR
APOSOR917812-1G
APOSOR917812-5G
7-Methoxyquinoline hydrochloride 95%
7-Methoxyquinoline hydrochloride
Formula:
C₁₀H₉NO·ClH MW: 195,646 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD22683321 Núm. CAS: 1418117-82-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...