Aminomethylboronic acid pinacol ester hydrochloride 95%
Proveedor: Apollo Scientific
OR919478-1G
OR919478-250MG
APOSOR919478-1GEA
284
EUR
APOSOR919478-1G
APOSOR919478-250MG
Aminomethylboronic acid pinacol ester hydrochloride 95%
Aminomethylboronic acid pinacol ester hydrochloride
Formula:
C₇H₁₆BNO₂·ClH Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD19288890 Núm. CAS: 298689-75-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...