(6-Chloropyridin-2-yl)methanamine dihydrochloride 95%

Proveedor: Apollo Scientific

OR919860-1G OR919860-250MG OR919860-5G
APOSOR919860-1GEA 151 EUR
APOSOR919860-1G APOSOR919860-250MG APOSOR919860-5G
(6-Chloropyridin-2-yl)methanamine dihydrochloride 95%
(6-Chloropyridin-2-yl)methanamine dihydrochloride
Formula: C₆H₇ClN₂·2ClH
MW: 215,508 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD23135891
Núm. CAS: 1557921-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR