(R)-Methyl 3-methylmorpholine-3-carboxylate hydrochloride 95%
Proveedor: Apollo Scientific
OR923430-1G
OR923430-250MG
APOSOR923430-1GEA
806
EUR
APOSOR923430-1G
APOSOR923430-250MG
(R)-Methyl 3-methylmorpholine-3-carboxylate hydrochloride 95%
(R)-Methyl 3-methylmorpholine-3-carboxylate hydrochloride
Formula:
C₇H₁₃NO₃·ClH Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD26406785 Núm. CAS: 1434126-90-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...