3-[(Ethylamino)methyl]phenol 95%
Proveedor: Apollo Scientific
OR926076-1G
OR926076-5G
APOSOR926076-1GEA
43.5
EUR
APOSOR926076-1G
APOSOR926076-5G
3-[(Ethylamino)methyl]phenol 95%
3-[(Ethylamino)methyl]phenol
Formula:
C₉H₁₃NO MW: 151,206 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD09931765 Núm. CAS: 91239-98-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...