5-Ethyl-2-hydroxybenzoic acid 95%
Proveedor: Apollo Scientific
OR926276-100MG
OR926276-1G
OR926276-250MG
APOSOR926276-100MGEA
161
EUR
APOSOR926276-100MG
APOSOR926276-1G
APOSOR926276-250MG
5-Ethyl-2-hydroxybenzoic acid 95%
5-Ethyl-2-hydroxybenzoic acid
Formula:
C₉H₁₀O₃ MW: 166,174 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD11203618 Núm. CAS: 51-27-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...