2-(Pyridin-2-yloxy)ethanamine 98%
Proveedor: Apollo Scientific
OR929033-1G
OR929033-5G
APOSOR929033-1GEA
178
EUR
APOSOR929033-1G
APOSOR929033-5G
2-(Pyridin-2-yloxy)ethanamine 98%
2-(Pyridin-2-yloxy)ethanamine
Formula:
C₇H₁₀N₂O MW: 138,167 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00090835 Núm. CAS: 29450-07-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...