tert-Butyl 2-(ethylamino)acetate 95%
Proveedor: Apollo Scientific
OR929179-1G
OR929179-250MG
APOSOR929179-1GEA
140
EUR
APOSOR929179-1G
APOSOR929179-250MG
tert-Butyl 2-(ethylamino)acetate 95%
tert-Butyl 2-(ethylamino)acetate
Formula:
C₈H₁₇NO₂ MW: 159,226 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD12148668 Núm. CAS: 172317-17-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...