6-Acetyl-7-hydroxytetralin 95%
Proveedor: Apollo Scientific
OR929629-1G
OR929629-250MG
APOSOR929629-1GEA
435
EUR
APOSOR929629-1G
APOSOR929629-250MG
6-Acetyl-7-hydroxytetralin 95%
6-Acetyl-7-hydroxytetralin
Formula:
C₁₂H₁₄O₂ MW: 190,238 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD06658290 Núm. CAS: 40420-05-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...