5,6,7,8-Tetrahydro-8-quinolinol acetate 95%
Proveedor: Apollo Scientific
OR929696-1G
OR929696-250MG
APOSOR929696-1GEA
507
EUR
APOSOR929696-1G
APOSOR929696-250MG
5,6,7,8-Tetrahydro-8-quinolinol acetate 95%
5,6,7,8-Tetrahydro-8-quinolinol acetate
Formula:
C₁₁H₁₃NO₂ MW: 191,226 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD11974482 Núm. CAS: 14631-47-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...