(1-Fmoc-aminocyclohexyl)acetic acid 97%
Proveedor: Apollo Scientific
OR930222-1G
APOSOR930222-1GEA
319
EUR
APOSOR930222-1G
(1-Fmoc-aminocyclohexyl)acetic acid 97%
(1-Fmoc-aminocyclohexyl)acetic acid
Formula:
C₂₃H₂₅NO₄ MW: 379,449 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD01863220 Núm. CAS: 282524-98-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...