N,N-Dimethyl-3-(2-pyrrolidinyl)aniline 95%
Proveedor: Apollo Scientific
OR931213-1G
OR931213-250MG
APOSOR931213-1GEA
806
EUR
APOSOR931213-1G
APOSOR931213-250MG
N,N-Dimethyl-3-(2-pyrrolidinyl)aniline 95%
N,N-Dimethyl-3-(2-pyrrolidinyl)aniline
Formula:
C₁₂H₁₈N₂ MW: 190,285 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD02663468 Núm. CAS: 383127-43-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...