Ethyl 6-chloropyrimidine-4-carboxylate 97%
Proveedor: Apollo Scientific
OR931255-25G
OR931255-5G
APOSOR931255-25GEA
388
EUR
APOSOR931255-25G
APOSOR931255-5G
Ethyl 6-chloropyrimidine-4-carboxylate 97%
Ethyl 6-chloropyrimidine-4-carboxylate
Formula:
C₇H₇ClN₂O₂ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD12756023 Núm. CAS: 51940-63-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...