3-[1-(Dimethylamino)ethyl]benzaldehyde 95%
Proveedor: Apollo Scientific
OR931608-1G
OR931608-250MG
APOSOR931608-1GEA
400
EUR
APOSOR931608-1G
APOSOR931608-250MG
3-[1-(Dimethylamino)ethyl]benzaldehyde 95%
3-[1-(Dimethylamino)ethyl]benzaldehyde
Formula:
C₁₁H₁₅NO MW: 177.243 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD08059975 Núm. CAS: 915924-91-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...