(S)-tert-Butyl (2-aminopropyl)carbamate 97%
Proveedor: Apollo Scientific
OR932018-1G
APOSOR932018-1GEA
272
EUR
APOSOR932018-1G
(S)-tert-Butyl (2-aminopropyl)carbamate 97%
(S)-tert-Butyl (2-aminopropyl)carbamate
Formula:
C₈H₁₈N₂O₂ MW: 174,241 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD11112235 Núm. CAS: 121103-15-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...