4-Chloro-6-methylnicotinic acid 95%
Proveedor: Apollo Scientific
OR932816-10G
OR932816-1G
OR932816-5G
APOSOR932816-10GEA
330
EUR
APOSOR932816-10G
APOSOR932816-1G
APOSOR932816-5G
4-Chloro-6-methylnicotinic acid 95%
4-Chloro-6-methylnicotinic acid
Formula:
C₇H₆ClNO₂ MW: 171,581 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD11520850 Núm. CAS: 1060805-95-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...