(R)-Benzyl 5-methyl-1,4-diazepane-1-carboxylate 95%
Proveedor: Apollo Scientific
OR942975-1G
OR942975-5G
APOSOR942975-1GEA
98.9
EUR
APOSOR942975-1G
APOSOR942975-5G
(R)-Benzyl 5-methyl-1,4-diazepane-1-carboxylate 95%
(R)-Benzyl 5-methyl-1,4-diazepane-1-carboxylate
Formula:
C₁₄H₂₀N₂O₂ MW: 248,321 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD22420241 Núm. CAS: 1001401-60-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...