Methyl 3-amino-5-ethoxybenzoate 95%
Proveedor: Apollo Scientific
OR950296-1G
APOSOR950296-1GEA
27.7
EUR
APOSOR950296-1G
Methyl 3-amino-5-ethoxybenzoate 95%
Methyl 3-amino-5-ethoxybenzoate
Formula:
C₁₀H₁₃NO₃ MW: 195,215 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD20696913 Núm. CAS: 706792-04-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...