N-(4-Ethoxybenzyl)-N-methylamine 98%
Proveedor: Apollo Scientific
OR952831-1G
OR952831-5G
APOSOR952831-1GEA
290
EUR
APOSOR952831-1G
APOSOR952831-5G
N-(4-Ethoxybenzyl)-N-methylamine 98%
N-(4-Ethoxybenzyl)-N-methylamine
Formula:
C₁₀H₁₅NO MW: 165.232 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD04622318 Núm. CAS: 41690-86-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...