Ethyl 4,5,6,7-tetrahydro-1H-indole-2-carboxylate 98%
Proveedor: Apollo Scientific
OR953421-1G
APOSOR953421-1GEA
209
EUR
APOSOR953421-1G
Ethyl 4,5,6,7-tetrahydro-1H-indole-2-carboxylate 98%
Ethyl 4,5,6,7-tetrahydro-1H-indole-2-carboxylate
Formula:
C₁₁H₁₅NO₂ MW: 193,242 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD14635837 Núm. CAS: 65880-18-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...