Methyl (3r)-3-amino-3-phenylpropanoate 95%

Proveedor: Apollo Scientific

OR955845-1G OR955845-5G
APOSOR955845-1GEA 190 EUR
APOSOR955845-1G APOSOR955845-5G
Methyl (3r)-3-amino-3-phenylpropanoate 95%
Methyl (3r)-3-amino-3-phenylpropanoate
Formula: C₁₀H₁₃NO₂
MW: 179,216 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD08056712
Núm. CAS: 37088-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR