2-(Isopropylamino)benzonitrile 98%
Proveedor: Apollo Scientific
OR959902-1G
OR959902-5G
APOSOR959902-1GEA
168
EUR
APOSOR959902-1G
APOSOR959902-5G
2-(Isopropylamino)benzonitrile 98%
2-(Isopropylamino)benzonitrile
Formula:
C₁₀H₁₂N₂ MW: 160,216 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD11144672 Núm. CAS: 147531-47-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...