N-(Thiophen-2-ylmethyl)cyclohexanamine 97%
Proveedor: Apollo Scientific
OR959967-1G
OR959967-5G
APOSOR959967-1GEA
219
EUR
APOSOR959967-1G
APOSOR959967-5G
N-(Thiophen-2-ylmethyl)cyclohexanamine 97%
N-(Thiophen-2-ylmethyl)cyclohexanamine
Formula:
C₁₁H₁₇NS MW: 195,324 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD04491291 Núm. CAS: 51305-86-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...