Methyl 1-isopropyl-2-oxopyrrolidine-4-carboxylate 95%
Proveedor: Apollo Scientific
OR965124-1G
OR965124-5G
APOSOR965124-1GEA
306
EUR
APOSOR965124-1G
APOSOR965124-5G
Methyl 1-isopropyl-2-oxopyrrolidine-4-carboxylate 95%
Methyl 1-isopropyl-2-oxopyrrolidine-4-carboxylate
Formula:
C₉H₁₅NO₃ MW: 185,22 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD11053005 Núm. CAS: 59857-84-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...