3-Pyrrolidin-1-ylacrylic acid methyl ester 95%

Proveedor: Apollo Scientific

OR965324-1G OR965324-25G OR965324-5G
APOSOR965324-1GEA 72.5 EUR
APOSOR965324-1G APOSOR965324-25G APOSOR965324-5G
3-Pyrrolidin-1-ylacrylic acid methyl ester 95%
3-Pyrrolidin-1-ylacrylic acid methyl ester
Formula: C₈H₁₃NO₂
MW: 155,194 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD01109140
Núm. CAS: 90087-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR