3-Phenylquinoxalin-2-(1H)-one 97%
Proveedor: Apollo Scientific
OR9871-1G
OR9871-5G
APOSOR9871-1GEA
183
EUR
APOSOR9871-1G
APOSOR9871-5G
3-Phenylquinoxalin-2-(1H)-one 97%
3-Phenylquinoxalin-2-(1H)-one
Formula:
C₁₄H₁₀N₂O MW: 222,242 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00798377 Núm. CAS: 1504-78-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...