(Triphenylphosphonio)difluoroacetate 95%
Proveedor: Apollo Scientific
Sinónimos:
2,2-Difluoro-2-(triphenylphosphonio)acetate
PC10699-5G
APOSPC10699-5GEA
102
EUR
APOSPC10699-5G
(Triphenylphosphonio)difluoroacetate 95%
(Triphenylphosphonio)difluoroacetate
Formula:
C₂₀H₁₅F₂O₂P MW: 356,302 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD28359690 Núm. CAS: 1449521-05-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...