3-Nitrobenzenesulphonyl fluoride 97%

Proveedor: Apollo Scientific

PC1787-25G PC1787-5G
APOSPC1787-25GEA 290 EUR
APOSPC1787-25G APOSPC1787-5G
3-Nitrobenzenesulphonyl fluoride 97%
3-Nitrobenzenesulphonyl fluoride
Formula: C₆H₄FNO₄S
MW: 205,164 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00024845
Núm. CAS: 349-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR