6-Fluoro-2,3-dihydro-1H-inden-1-amine hydrochloride 98%
Proveedor: Apollo Scientific
PC201045-100MG
PC201045-1G
PC201045-250MG
APOSPC201045-100MGEA
96
EUR
APOSPC201045-100MG
APOSPC201045-1G
APOSPC201045-250MG
6-Fluoro-2,3-dihydro-1H-inden-1-amine hydrochloride 98%
6-Fluoro-2,3-dihydro-1H-inden-1-amine hydrochloride
Formula:
C₉H₁₀FN·ClH MW: 187,642 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD08544167 Núm. CAS: 1191908-44-7 |
Learn more
![](/m-es.vwr.com/es_ES/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...