Methyl 6-(fluoromethyl)picolinate 97%

Proveedor: Apollo Scientific

PC201056-100MG PC201056-1G PC201056-250MG
APOSPC201056-100MGEA 104 EUR
APOSPC201056-100MG APOSPC201056-1G APOSPC201056-250MG
Methyl 6-(fluoromethyl)picolinate 97%
Methyl 6-(fluoromethyl)picolinate
Formula: C₈H₈FNO₂
MW: 169,153 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD11109900
Núm. CAS: 1209248-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR