4-Bromo-2-chloro-3-fluoropyridine 95+%
Proveedor: Apollo Scientific
PC201060-100MG
PC201060-1G
PC201060-250MG
APOSPC201060-100MGEA
105
EUR
APOSPC201060-100MG
APOSPC201060-1G
APOSPC201060-250MG
4-Bromo-2-chloro-3-fluoropyridine 95+%
4-Bromo-2-chloro-3-fluoropyridine
Formula:
C₅H₂BrClFN MW: 210,431 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD16606306 Núm. CAS: 1211526-56-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...