(R)-2-(4-Fluorophenyl)pyrrolidine hydrochloride 95+%

Proveedor: Apollo Scientific

PC201131-100MG PC201131-250MG
APOSPC201131-100MGEA 40.6 EUR
APOSPC201131-100MG APOSPC201131-250MG
(R)-2-(4-Fluorophenyl)pyrrolidine hydrochloride 95+%
(R)-2-(4-Fluorophenyl)pyrrolidine hydrochloride
Formula: C₁₀H₁₂FN·ClH
Temperatura de almacenaje: Ambiente
Número MDL: MFCD08751397
Núm. CAS: 1381928-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR