5-Fluoro-2-hydrazinylpyridine dihydrochloride 95+%

Proveedor: Apollo Scientific

PC201133-1G PC201133-250MG PC201133-5G
APOSPC201133-1GEA 144 EUR
APOSPC201133-1G APOSPC201133-250MG APOSPC201133-5G
5-Fluoro-2-hydrazinylpyridine dihydrochloride 95+%
5-Fluoro-2-hydrazinylpyridine dihydrochloride
Formula: C₅H₆FN₃·2ClH
MW: 200,042 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD20441448
Núm. CAS: 1401426-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR