2-Fluorocyclopropanecarboxylic acid 97%
Proveedor: Apollo Scientific
PC201153-1G
APOSPC201153-1GEA
60.9
EUR
APOSPC201153-1G
2-Fluorocyclopropanecarboxylic acid 97%
2-Fluorocyclopropanecarboxylic acid
Formula:
C₄H₅FO₂ MW: 104,08 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD21604147 Núm. CAS: 156816-78-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...