2-Fluoro-3-(hydroxymethyl)benzoic acid
Proveedor: Apollo Scientific
PC2643-1G
PC2643-250MG
PC2643-5G
APOSPC2643-1GEA
637
EUR
APOSPC2643-1G
APOSPC2643-250MG
APOSPC2643-5G
2-Fluoro-3-(hydroxymethyl)benzoic acid
2-Fluoro-3-(hydroxymethyl)benzoic acid
Formula:
C₈H₇FO₃ MW: 170,138 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD01862008 Núm. CAS: 481075-37-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...