2-{[(9H-Fluoren-9-ylmethoxy)carbonyl]amino}-2-[2-(trifluoromethoxy)phenyl]acetic acid
Proveedor: Apollo Scientific
PC27859-100MG
PC27859-1G
PC27859-250MG
APOSPC27859-100MGEA
633
EUR
APOSPC27859-100MG
APOSPC27859-1G
APOSPC27859-250MG
2-{[(9H-Fluoren-9-ylmethoxy)carbonyl]amino}-2-[2-(trifluoromethoxy)phenyl]acetic acid
2-{[(9H-Fluoren-9-ylmethoxy)carbonyl]amino}-2-[2-(trifluoromethoxy)phenyl]acetic acid
Formula:
C₂₄H₁₈F₃NO₅ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD28389401 Núm. CAS: 1692823-37-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...